MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Transcription suppression is mediated by the HDAC1-Sin3 complex in Xenopus nucleoplasmic extract.
Transcription suppression is mediated by the HDAC1-Sin3 complex in Xenopus nucleoplasmic extract.
The Journal of biological chemistry (2022-10-12)
Colleen E Quaas, Baicheng Lin, David T Long
ABSTRACT
Modification of histones provides a dynamic mechanism to regulate chromatin structure and access to DNA. Histone acetylation, in particular, plays a prominent role in controlling the interaction between DNA, histones, and other chromatin-associated proteins. Defects in histone acetylation patterns interfere with normal gene expression and underlie a wide range of human diseases. Here, we utilize Xenopus egg extracts to investigate how changes in histone acetylation influence transcription of a defined gene construct. We show that inhibition of histone deacetylase 1 and 2 (HDAC1/2) specifically counteracts transcription suppression by preventing chromatin compaction and deacetylation of histone residues H4K5 and H4K8. Acetylation of these sites supports binding of the chromatin reader and transcription regulator BRD4. We also identify HDAC1 as the primary driver of transcription suppression and show that this activity is mediated through the Sin3 histone deacetylase complex. These findings highlight functional differences between HDAC1 and HDAC2, which are often considered to be functionally redundant, and provide additional molecular context for their activity.
Sigma-Aldrich
Anti-CoREST Antibody, Upstate®, from rabbit
Sigma-Aldrich
(+)-JQ1, ≥98% (HPLC)