MilliporeSigma
  • High glucose upregulates PAR-1 in SH-SY5Y cells via deficiency of miR-20a and miR-190a.
High glucose upregulates PAR-1 in SH-SY5Y cells via deficiency of miR-20a and miR-190a.
Fundamental & clinical pharmacology (2021-12-15)
Li Kong, Pan-Pan Gu, Zhuang-Zhuang Tang, Ling-Shan Gou, Yao-Wu Liu
ABSTRACT
Thrombin activity enhancement and its receptor protease-activated receptor 1 (PAR-1) activation play vital roles in neurologic deficits in the central nervous system. Our recent study showed that PAR-1 upregulation stimulated by chronic high glucose (HG) caused central neuron injury through neuroinflammation; however, the molecular mechanisms are far from clear. In the present study, we found that HG resulted in neuronal injury of SH-SY5Y cells as evidenced by decreased cell viability and increased lactate dehydrogenase release and elevated the mRNA level of PAR-1. Moreover, we predicted and determined several potential microRNAs (miRs) combining with the 3'-UTR of PAR-1 mRNA, finding that miR-20a-5p, miR-93-5p, and miR-190a-5p were significantly decreased in HG-cultured SH-SY5Y cells compared with control. Further, SH-SY5Y cells stably transfected with miR-20a-5p or miR-190a-5p mimic were established, and overexpression efficiency were confirmed. It was found that miR-20a-5p or miR-190a-5p overexpression markedly decreased PAR-1 mRNA level and protein expression in SH-SY5Y cells cultured with HG and normal glucose, indicating that miR-20a or miR-19a deficiency contributed to HG-induced PAR-1 upregulation. Together, our findings demonstrated that PAR-1 upregulation mediated HG-induced neuronal damage in central neurons, which was achieved through miR-20a or miR-190a deficiency.
Sigma-Aldrich
Anti-PAR1 (Cleaved-Ser42), N-Terminal antibody produced in rabbit, affinity isolated antibody