MilliporeSigma
  • Role of mitogen-activated protein kinases, NF-kappaB, and AP-1 on cerulein-induced IL-8 expression in pancreatic acinar cells.
Role of mitogen-activated protein kinases, NF-kappaB, and AP-1 on cerulein-induced IL-8 expression in pancreatic acinar cells.
Annals of the New York Academy of Sciences (2007-03-27)
Kyung Don Ju, Ji Hoon Yu, Hyeyoung Kim, Kyung Hwan Kim
ABSTRACT
The cholecystokine (CCK) analogue cerulein causes pathophysiological, morphological, and biochemical events similar to various aspects of human pancreatitis. Doses of CCK or cerulein beyond those that cause the maximum pancreatic secretion of amylase and lipase result in pancreatitis, which is characterized by a dysregulation of the digestive enzyme production and cytoplasmic vacuolization and the death of acinar cells, edema formation, and an infiltration of inflammatory cells into the pancreas. This study aims to investigate whether cerulein induces IL-8 expression in pancreatic acinar cells, and whether cerulein-induced IL-8 expression is inhibited in the cells transfected with mutant genes for c-jun (TAM-67), or IkappaBalpha (MAD-3) or treated inhibitors of mitogen-activated protein kinases (MAPKs). As a result, cerulein induced IL-expression, which was inhibited in the cells transfected with TAM-67 or MAD-3 or treated inhibitors of MAPK. In conclusion, activation of MAPK, nuclear factor-kappaB (NF-kappaB), and activator protein-1 (AP-1) may be the upstream signaling for cerulein-induced IL-8 expression in pancreatic acinar cells.
Sigma-Aldrich
SB 203580, SB 203580, CAS 152121-47-6, is a highly specific, potent, cell-permeable, selective, reversible, and ATP-competitive inhibitor of p38 MAP kinase (IC50 = 34 nM in vitro, 600 nM in cells).