MilliporeSigma
  • Mechanistic investigations of the liver toxicity of the free fatty acid receptor 1 agonist fasiglifam (TAK875) and its primary metabolites.
Mechanistic investigations of the liver toxicity of the free fatty acid receptor 1 agonist fasiglifam (TAK875) and its primary metabolites.
Journal of biochemical and molecular toxicology (2019-05-09)
Timothy Ackerson, Alexander Amberg, Jens Atzrodt, Catherine Arabeyre, Elisabeth Defossa, Martina Dorau, Angela Dudda, Jacquelyn Dwyer, Wolfgang Holla, Thomas Kissner, Markus Kohlmann, Ulrich Kürzel, József Pánczél, Shibani Rajanna, Jens Riedel, Friedemann Schmidt, Kerstin Wäse, Dietmar Weitz, Volker Derdau
ABSTRACT
For fasiglifam (TAK875) and its metabolites the substance-specific mechanisms of liver toxicity were studied. Metabolism studies were run to identify a putatively reactive acyl glucuronide metabolite. In vitro cytotoxicity and caspase 3/7 activation were assessed in primary human and dog hepatocytes in 2D and 3D cell culture. Involvement of glutathione (GSH) detoxication system in mediating cytotoxicity was determined by assessing potentiation of cytotoxicity in a GSH depleted in vitro system. In addition, potential mitochondrial liabilities of the compounds were assessed in a whole-cell mitochondrial functional assay. Fasiglifam showed moderate cytotoxicity in human primary hepatocytes in the classical 2D cytotoxicity assays and also in the complex 3D human liver microtissue (hLiMT) after short-term treatment (24 hours or 48 hours) with TC50 values of 56 to 68 µM (adenosine triphosphate endpoint). The long-term treatment for 14 days in the hLiMT resulted in a slight TC50 shift over time of 2.7/3.6 fold lower vs 24-hour treatment indicating possibly a higher risk for cytotoxicity during long-term treatment. Cellular GSH depletion and impairment of mitochondrial function by TAK875 and its metabolites evaluated by Seahorse assay could not be found being involved in DILI reported for TAK875. The acyl glucuronide metabolites of TAK875 have been finally identified to be the dominant reason for liver toxicity.
Sigma-Aldrich
L-Buthionine-sulfoximine, ≥97% (TLC)