MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • GSK3α phosphorylates dynamin-2 to promote GLUT4 endocytosis in muscle cells.
GSK3α phosphorylates dynamin-2 to promote GLUT4 endocytosis in muscle cells.
The Journal of cell biology (2022-11-30)
Jessica Laiman, Yen-Jung Hsu, Julie Loh, Wei-Chun Tang, Mei-Chun Chuang, Hui-Kang Liu, Wei-Shun Yang, Bi-Chang Chen, Lee-Ming Chuang, Yi-Cheng Chang, Ya-Wen Liu
ABSTRACT
Insulin-stimulated translocation of glucose transporter 4 (GLUT4) to plasma membrane of skeletal muscle is critical for postprandial glucose uptake; however, whether the internalization of GLUT4 is also regulated by insulin signaling remains unclear. Here, we discover that the activity of dynamin-2 (Dyn2) in catalyzing GLUT4 endocytosis is negatively regulated by insulin signaling in muscle cells. Mechanistically, the fission activity of Dyn2 is inhibited by binding with the SH3 domain of Bin1. In the absence of insulin, GSK3α phosphorylates Dyn2 to relieve the inhibition of Bin1 and promotes endocytosis. Conversely, insulin signaling inactivates GSK3α and leads to attenuated GLUT4 internalization. Furthermore, the isoform-specific pharmacological inhibition of GSK3α significantly improves insulin sensitivity and glucose tolerance in diet-induced insulin-resistant mice. Together, we identify a new role of GSK3α in insulin-stimulated glucose disposal by regulating Dyn2-mediated GLUT4 endocytosis in muscle cells. These results highlight the isoform-specific function of GSK3α on membrane trafficking and its potential as a therapeutic target for metabolic disorders.
Product Description
Millipore
Monoclonal Anti-HA−Agarose antibody produced in mouse, clone HA-7, purified immunoglobulin, PBS suspension