MilliporeSigma
  • Neuronal calcium channel antagonists. Discrimination between calcium channel subtypes using omega-conotoxin from Conus magus venom.
Neuronal calcium channel antagonists. Discrimination between calcium channel subtypes using omega-conotoxin from Conus magus venom.
Biochemistry (1987-04-21)
B M Olivera, L J Cruz, V de Santos, G W LeCheminant, D Griffin, R Zeikus, J M McIntosh, R Galyean, J Varga, W R Gray
ABSTRACT
The omega-conotoxins from the venom of fish-hunting cone snails are probably the most useful of presently available ligands for neuronal Ca channels from vertebrates. Two of these peptide toxins, omega-conotoxins MVIIA and MVIIB from the venom of Conus magus, were purified. The amino acid sequences show significant differences from omega-conotoxins from Conus geographus. Total synthesis of omega-conotoxin MVIIA was achieved, and biologically active radiolabeled toxin was produced by iodination. Although omega-conotoxins from C. geographus (GVIA) and C. magus (MVIIA) appear to compete for the same sites in mammalian brain, in amphibian brain the high-affinity binding of omega-conotoxin MVIIA has narrower specificity. In this system, it is demonstrated that a combination of two omega-conotoxins can be used for biochemically defining receptor subtypes and suggested that these correspond to subtypes of neuronal Ca2+ channels.