MilliporeSigma
  • Preparation of silver nanoparticles and their industrial and biomedical applications: a comprehensive review
Preparation of silver nanoparticles and their industrial and biomedical applications: a comprehensive review
Advances in Materials Science and Engineering (2015)
Adnan Haider and Inn-Kyu Kang