MilliporeSigma
  • Combined time-restricted feeding and cisplatin enhance the anti-tumor effects in cisplatin-resistant and -sensitive lung cancer cells.
Combined time-restricted feeding and cisplatin enhance the anti-tumor effects in cisplatin-resistant and -sensitive lung cancer cells.
Medical oncology (Northwood, London, England) (2022-12-29)
Jianling Li, Qianyao Chen, Dan Shi, Xuemei Lian
ABSTRACT
Combination therapy as an important treatment option for lung cancer has been attracting attention due to the primary and acquired resistance of chemotherapeutic drugs in the clinical application. In the present study, as a new therapy strategy, concomitant treatment with time-restricted feeding (TRF) plus cisplatin (DDP) on lung cancer growth was investigated in DDP-resistant and DDP-sensitive lung cancer cells. We first found that TRF significantly enhanced the drug susceptibility of DDP in DDP-resistant A549 (A549/DDP) cell line, illustrated by reversing the inhibitory concentration 50 (IC50) values of A549/DDP cells to normal level of parental A549 cells. We also found that TRF markedly enhanced DDP inhibition on cell proliferation, migration, as well as promoted apoptosis compared to the DDP alone group in A549, H460 and A549/DDP cells lines. We further revealed that the synergistic anti-tumor effect of combined DDP and TRF was greater than that of combined DDP and simulated fasting condition (STS), a known anti-tumor cellular medium. Moreover, mRNA sequence analysis from A549/DDP cell line demonstrated the synergistic anti-tumor effect involved in upregulated pathways in p53 signaling pathway and apoptosis. Notably, compared with the DDP alone group, combination of TRF and DDP robustly upregulated the P53 protein expression without mRNA level change by regulating its stability via promoting protein synthesis and inhibiting degradation, revealed by cycloheximide and MG132 experiments. Collectively, our results suggested that TRF in combination with cisplatin might be an additional novel therapeutic strategy for patients with lung cancer.
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline