MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Proteome profiling of cerebrospinal fluid reveals biomarker candidates for Parkinson's disease.
Proteome profiling of cerebrospinal fluid reveals biomarker candidates for Parkinson's disease.
Cell reports. Medicine (2022-06-23)
Ozge Karayel, Sebastian Virreira Winter, Shalini Padmanabhan, Yuliya I Kuras, Duc Tung Vu, Idil Tuncali, Kalpana Merchant, Anne-Marie Wills, Clemens R Scherzer, Matthias Mann
ABSTRACT
Parkinson's disease (PD) is a growing burden worldwide, and there is no reliable biomarker used in clinical routines to date. Cerebrospinal fluid (CSF) is routinely collected in patients with neurological symptoms and should closely reflect alterations in PD patients' brains. Here, we describe a scalable and sensitive mass spectrometry (MS)-based proteomics workflow for CSF proteome profiling. From two independent cohorts with over 200 individuals, our workflow reproducibly quantifies over 1,700 proteins from minimal CSF amounts. Machine learning determines OMD, CD44, VGF, PRL, and MAN2B1 to be altered in PD patients or to significantly correlate with clinical scores. We also uncover signatures of enhanced neuroinflammation in LRRK2 G2019S carriers, as indicated by increased levels of CTSS, PLD4, and HLA proteins. A comparison with our previously acquired urinary proteomes reveals a large overlap in PD-associated changes, including lysosomal proteins, opening up new avenues to improve our understanding of PD pathogenesis.
Sigma-Aldrich
Trypsin from porcine pancreas, Proteomics Grade, BioReagent, Dimethylated