MilliporeSigma
  • Primary structure of bovine brain acidic fibroblast growth factor (FGF).
Primary structure of bovine brain acidic fibroblast growth factor (FGF).
Biochemical and biophysical research communications (1985-12-17)
F Esch, N Ueno, A Baird, F Hill, L Denoroy, N Ling, D Gospodarowicz, R Guillemin
ABSTRACT
The major anionic mitogenic polypeptide for endothelial cells, acidic fibroblast growth factor (FGF), has been purified to homogeneity from bovine brain and its complete primary structure established by gas-phase sequence analysis. The 140 amino acid (Mr 16,000) protein has been previously shown to be a potent growth factor for many diverse cell types of mesodermal origin, in vitro, and an angiogenic agent, in vivo. The amino acid sequence of bovine brain acidic FGF has a 53% absolute homology with that of bovine pituitary basic FGF suggesting that these endothelial cell mitogens are derived from a single ancestral gene.