MilliporeSigma
  • Synthetic peptide derivatives that bind to fibrinogen and prevent the polymerization of fibrin monomers.
Synthetic peptide derivatives that bind to fibrinogen and prevent the polymerization of fibrin monomers.
Proceedings of the National Academy of Sciences of the United States of America (1978-07-01)
A P Laudano, R F Doolittle
PMID277910
ABSTRACT
A series of small peptides corresponding to the amino termini of the fibrin alpha- and beta-chains has been synthesized. The peptides glycyl-L-prolyl-L-arginyl-L-proline and glycyl-L-prolyl-L-arginylsarcosine are potent inhibitors of fibrin polymerization. Moreover, these peptides have a natural stability stemming from their inherent resistance to proteolysis because of the involvement of amino acids in each of their peptide bonds. The peptide glycyl-L-prolyl-L-arginyl-L-proline binds to fibrinogen and to fragment D, in both cases with an association constant of approximately 5 x 10(4); it does not bind to fragment E. The number of binding sites is two for fibrinogen and one for fragment D. The tripeptide glycyl-L-prolyl-L-arginine binds less tightly and is less than half as effective in preventing polymerization. The peptide glycyl-L-histidyl-L-arginyl-L-proline, which corresponds exactly to the amino terminus of the fibrin beta-chain, does not inhibit the aggregation of fibrin monomers under the conditions used. It does bind weakly to fibrinogen, however, suggesting the involvement of sites other than those binding the alpha-chain analogues. Various other peptides were found not to inhibit polymerization; these included glycine-L-proline, L-prolyl-L-arginine and glycyl-L-prolyl-L-seryl-L-proline. The last-named corresponds to the serine/arginine amino acid replacement previously reported for a defective human fibrinogen.