MilliporeSigma
  • P97/VCP ATPase inhibitors can rescue p97 mutation-linked motor neuron degeneration.
P97/VCP ATPase inhibitors can rescue p97 mutation-linked motor neuron degeneration.
Brain communications (2022-07-23)
F Wang, S Li, T Y Wang, G A Lopez, I Antoshechkin, T F Chou
ABSTRACT
Mutations in p97/VCP cause two motor neuron diseases: inclusion body myopathy associated with Paget disease of bone and frontotemporal dementia and familial amyotrophic lateral sclerosis. How p97 mutations lead to motor neuron degeneration is, however, unknown. Here we used patient-derived induced pluripotent stem cells to generate p97 mutant motor neurons. We reduced the genetic background variation by comparing mutant motor neurons to its isogenic wild type lines. Proteomic analysis reveals that p97R155H/+ motor neurons upregulate several cell cycle proteins at Day 14, but this effect diminishes by Day 20. Molecular changes linked to delayed cell cycle exit are observed in p97 mutant motor neurons. We also find that two p97 inhibitors, CB-5083 and NMS-873, restore some dysregulated protein levels. In addition, two p97 inhibitors and a food and drug administration-approved cyclin-dependent kinase 4/6 inhibitor, Abemaciclib, can rescue motor neuron death. Overall, we successfully used iPSC-derived motor neurons, identified dysregulated proteome and transcriptome and showed that p97 inhibitors rescue phenotypes in this disease model.
Sigma-Aldrich
Y-27632 dihydrochloride, ≥98% (HPLC)
BrainFast Motor Supplement, Neuronal Maturation Supplement (1000x)