MilliporeSigma
  • Purification of hetero-oligomeric protein variants using a modified tandem affinity purification approach.
Purification of hetero-oligomeric protein variants using a modified tandem affinity purification approach.
STAR protocols (2023-03-01)
Aasna L Parui, Kakoli Bose
ABSTRACT
Here, we describe a protocol for artificially generating hetero-oligomeric protein complexes from the homo-oligomers using a sequential denaturation-renaturation strategy, followed by a modified affinity chromatography protocol used for their purification. This protocol enables one to obtain a homogenous population of hetero-oligomers and understand the contribution of each protomer through further biochemical and/or biophysical characterization. For complete details on the use and execution of this protocol, please refer to Parui et al. (2022).1.
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)