MilliporeSigma
  • Effects of Suramin on Polycystic Kidney Disease in a Mouse Model of Polycystin-1 Deficiency.
Effects of Suramin on Polycystic Kidney Disease in a Mouse Model of Polycystin-1 Deficiency.
International journal of molecular sciences (2022-08-13)
Ming-Yang Chang, Shen-Hsing Hsu, Li-Yi Ma, Li-Feng Chou, Cheng-Chieh Hung, Ya-Chung Tian, Chih-Wei Yang
ABSTRACT
The aberrant activation of the purinergic signaling pathway has been shown to promote cyst growth and fluid secretion in autosomal dominant polycystic kidney disease (ADPKD). Suramin is an anti-parasitic drug that has strong anti-purinergic properties. Whether suramin could have a therapeutic effect on ADPKD has not been fully investigated. We examined the effect of suramin on cyst progression in a Pkd1 microRNAs transgenic mouse model that presented stable Pkd1 knockdown and moderate disease progression. The Pkd1-deficient mice were treated with suramin (60 mg/kg) by intraperitoneal injection twice a week from postnatal days 35 to 90. Kidney-to-body weight ratios, cyst indices, and blood urea nitrogen (BUN) levels were measured. Cell proliferation and macrophage infiltration were determined by immunohistochemistry. The suramin-treated group had significantly lower renal cyst densities, cell proliferation, and macrophage infiltration compared with saline-treated controls. Suramin significantly inhibited ERK phosphorylation and the expression of Il1b, Il6, Nlrp3, Tgfb, Fn1, P2rx7, and P2ry2 mRNAs in the kidneys. However, BUN levels remained high despite the reduction in cyst growth. Furthermore, plasma cystatin C and neutrophil gelatinase-associated lipocalin (NGAL) levels were significantly higher in the suramin-treated group compared with the control group. Periodic acid-Schiff staining revealed degenerative changes and epithelial cell vacuolation in the non-cystic renal tubules, which indicated phospholipidosis following suramin treatment. These results suggest that suramin may reduce renal cyst growth and inflammation, but the associated tubular cell injuries could limit its therapeutic potential. Other purinergic receptor antagonists with less nephrotoxicity may deserve further investigation for the treatment of ADPKD.
Sigma-Aldrich
Goat Anti-Rat light chain Antibody, biotin conjugate, Chemicon®, from goat