MilliporeSigma
Modeling human skeletal development using human pluripotent stem cells.
Proceedings of the National Academy of Sciences of the United States of America (2023-05-01)
Shireen R Lamandé, Elizabeth S Ng, Trevor L Cameron, Louise H W Kung, Lisa Sampurno, Lynn Rowley, Jinia Lilianty, Yudha Nur Patria, Tayla Stenta, Eric Hanssen, Katrina M Bell, Ritika Saxena, Kathryn S Stok, Edouard G Stanley, Andrew G Elefanty, John F Bateman
ABSTRACT
Chondrocytes and osteoblasts differentiated from induced pluripotent stem cells (iPSCs) will provide insights into skeletal development and genetic skeletal disorders and will generate cells for regenerative medicine applications. Here, we describe a method that directs iPSC-derived sclerotome to chondroprogenitors in 3D pellet culture then to articular chondrocytes or, alternatively, along the growth plate cartilage pathway to become hypertrophic chondrocytes that can transition to osteoblasts. Osteogenic organoids deposit and mineralize a collagen I extracellular matrix (ECM), mirroring in vivo endochondral bone formation. We have identified gene expression signatures at key developmental stages including chondrocyte maturation, hypertrophy, and transition to osteoblasts and show that this system can be used to model genetic cartilage and bone disorders.
Sigma-Aldrich
Anti-Lubricin Antibody/Proteoglycan 4, clone 9G3, clone 9G3, from mouse
Sigma-Aldrich
Anti-Collagen Type II Antibody, clone 6B3, clone 6B3, Chemicon®, from mouse