MilliporeSigma
  • Placental Streptococcus agalactiae DNA is associated with neonatal unit admission and foetal pro-inflammatory cytokines in term infants.
Placental Streptococcus agalactiae DNA is associated with neonatal unit admission and foetal pro-inflammatory cytokines in term infants.
Nature microbiology (2023-11-30)
Francesca Gaccioli, Katie Stephens, Ulla Sovio, Flora Jessop, Hilary S Wong, Susanne Lager, Emma Cook, Marcus C de Goffau, Kirsty Le Doare, Sharon J Peacock, Julian Parkhill, D Stephen Charnock-Jones, Gordon C S Smith
ABSTRACT
Streptococcus agalactiae (Group B Streptococcus; GBS) is a common cause of sepsis in neonates. Previous work detected GBS DNA in the placenta in ~5% of women before the onset of labour, but the clinical significance of this finding is unknown. Here we re-analysed this dataset as a case control study of neonatal unit (NNU) admission. Of 436 infants born at term (≥37 weeks of gestation), 7/30 with placental GBS and 34/406 without placental GBS were admitted to the NNU (odds ratio (OR) 3.3, 95% confidence interval (CI) 1.3-7.8). We then performed a validation study using non-overlapping subjects from the same cohort. This included a further 239 cases of term NNU admission and 686 term controls: 16/36 with placental GBS and 223/889 without GBS were admitted to the NNU (OR 2.4, 95% CI 1.2-4.6). Of the 36 infants with placental GBS, 10 were admitted to the NNU with evidence of probable but culture-negative sepsis (OR 4.8, 95% CI 2.2-10.3), 2 were admitted with proven GBS sepsis (OR 66.6, 95% CI 7.3-963.7), 6 were admitted and had chorioamnionitis (inflammation of the foetal membranes) (OR 5.3, 95% CI 2.0-13.4), and 5 were admitted and had funisitis (inflammation of the umbilical cord) (OR 6.7, 95% CI 12.5-17.7). Foetal cytokine storm (two or more pro-inflammatory cytokines >10 times median control levels in umbilical cord blood) was present in 36% of infants with placental GBS DNA and 4% of cases where the placenta was negative (OR 14.2, 95% CI 3.6-60.8). Overall, ~1 in 200 term births had GBS detected in the placenta, which was associated with infant NNU admission and morbidity.
Product Description
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks