MilliporeSigma
  • Notch Signaling Regulates the Chondrogenic Potential of Both Articular Chondrocytes and Their Progenitors During Expansion.
Notch Signaling Regulates the Chondrogenic Potential of Both Articular Chondrocytes and Their Progenitors During Expansion.
Stem cells (Dayton, Ohio) (2023-04-22)
Anastasiia D Kurenkova, Lei Li, Anna P Usanova, Xiaogang Feng, Baoyi Zhou, Andrei A Nedorubov, Alexey V Lychagin, Andrei S Chagin
ABSTRACT
Articular cartilage has a limited capacity for self-repair and clinical approaches to cartilage regeneration are needed. The only such approach developed to date involves an expansion of primary autologous chondrocytes in culture, followed by their reimplantation into a cartilage defect. However, because of the formation of fibrocartilage instead of hyaline cartilage, the outcome is often not satisfactory. It happens due to the de-differentiation of chondrocytes during the expansion step. Indeed, articular chondrocytes are non-proliferative and require partial or complete dedifferentiation before actively proliferating. In recent years stem/progenitor cells in articular cartilage (artSPCs) have been described. These cells maintain their own population and renew articular cartilage in sexually mature mice. artSPCs can, theoretically, be superior to chondrocytes, for repairing damaged cartilage. Accordingly, here, we searched for conditions that allow rapid expansion of both artSPCs and chondrocytes with simultaneous preservation of their ability to form hyaline cartilage. Among the modulators of Wnt, Notch, and FGF signaling and of cell adhesion screened, only fibronectin and modulators of the Notch pathway promoted the rapid expansion of artSPCs. Surprisingly, both inhibition and activation of the pathway had this effect. However, only inhibition of Notch during expansion facilitated the chondrogenic potential of both artSPCs and primary chondrocytes, whereas activation of this pathway abrogated this potential entirely. This effect was the same for murine and human cells. Our present observations indicate that Notch signaling is the major regulator of the chondrogenic capacity of both artSPCs and chondrocytes during their expansion.
Sigma-Aldrich
Anti-SOX9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Collagen I antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-MEF2C antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution