MilliporeSigma
  • Identification of novel biallelic LRRC6 variants in male Chinese patients with primary ciliary dyskinesia and infertility.
Identification of novel biallelic LRRC6 variants in male Chinese patients with primary ciliary dyskinesia and infertility.
Journal of assisted reproduction and genetics (2022-12-15)
Yunhao Li, Yong Li, Ying Wang, Lanlan Meng, Chen Tan, Juan Du, Yue-Qiu Tan, Hongchuan Nie, Qianjun Zhang, Guangxiu Lu, Ge Lin, Huanzhu Li, Huan Zhang, Chaofeng Tu
ABSTRACT
The aim of this study is to identify the genetic cause of primary ciliary dyskinesia (PCD) and male infertility in two unrelated Han Chinese families. We performed whole-exome sequencing in two unrelated male Han Chinese patients suffering from infertility and PCD to identify the pathogenic variants. Ultrastructural and immunostaining analyses of patient's spermatozoa were performed to characterize the effect of the variants. The pathogenicity of the variants was validated using patient's spermatozoa by western blotting and immunostaining analysis. Intracytoplasmic sperm injection (ICSI) was conducted in the affected families. Three variants in leucine-rich repeat containing 6 (LRRC6) [patient 1(compound heterozygote): NM_012472: c.538C > T, (p.R180*) and c.64dupT, (p.S22Ffs*19); patient 2 (homozygote): c.863C > A, (p.P288H)] were identified in two unrelated patients with PCD and male infertility. These variants were predicated deleterious and were absent or rare in human population genome data. LRRC6-mutant spermatozoa showed a highly aberrant morphology and ultrastructure with lacked inner and outer dynein arms. The LRRC6 protein was present along the normal sperm flagella, and was significantly decreased in the mutated spermatozoa. Interestingly, both patients were able to conceive through ICSI and birthed a healthy baby. Our results extend the LRRC6 variant spectrum and provide reproductive guidance to families suffering from PCD-linked infertility caused by LRRC6 variants.
Sigma-Aldrich
Anti-LRRC6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2