MilliporeSigma
  • Uncovering the gastrointestinal passage, intestinal epithelial cellular uptake, and AGO2 loading of milk miRNAs in neonates using xenomiRs as tracers.
Uncovering the gastrointestinal passage, intestinal epithelial cellular uptake, and AGO2 loading of milk miRNAs in neonates using xenomiRs as tracers.
The American journal of clinical nutrition (2023-03-25)
Patrick Philipp Weil, Susanna Reincke, Christian Alexander Hirsch, Federica Giachero, Malik Aydin, Jonas Scholz, Franziska Jönsson, Claudia Hagedorn, Duc Ninh Nguyen, Thomas Thymann, Anton Pembaur, Valerie Orth, Victoria Wünsche, Ping-Ping Jiang, Stefan Wirth, Andreas C W Jenke, Per Torp Sangild, Florian Kreppel, Jan Postberg
ABSTRACT
Human breast milk has a high microRNA (miRNA) content. It remains unknown whether and how milk miRNAs might affect intestinal gene regulation and homeostasis of the developing microbiome after initiating enteral nutrition. However, this requires that relevant milk miRNA amounts survive the gastrointestinal (GI) passage, are taken up by cells, and become available to the RNA interference machinery. It seems important to dissect the fate of these miRNAs after oral ingestion and GI passage. Our goal was to analyze the potential transmissibility of milk miRNAs via the gastrointestinal system in neonate humans and a porcine model in vivo to contribute to the discussion of whether milk miRNAs could influence gene regulation in neonates and thus might vertically transmit developmental relevant signals. We performed cross-species profiling of miRNAs via deep sequencing and utilized dietary xenobiotic taxon-specific milk miRNA (xenomiRs) as tracers in human and porcine neonates, followed by functional studies in primary human fetal intestinal epithelial cells using adenovirus-type 5-mediated miRNA gene transfer. Mammals share many milk miRNAs yet exhibit taxon-specific miRNA fingerprints. We traced bovine-specific miRNAs from formula nutrition in human preterm stool and 9 d after the onset of enteral feeding in intestinal cells (ICs) of preterm piglets. Thereafter, several xenomiRs accumulated in the ICs. Moreover, a few hours after introducing enteral feeding in preterm piglets with supplemented reporter miRNAs (cel-miR-39-5p/-3p), we observed their enrichment in blood serum and in argonaute RISC catalytic component 2 (AGO2)-immunocomplexes from intestinal biopsies. Milk-derived miRNAs survived GI passage in human and porcine neonates. Bovine-specific miRNAs accumulated in ICs of preterm piglets after enteral feeding with bovine colostrum/formula. In piglets, colostrum supplementation with cel-miR-39-5p/-3p resulted in increased blood concentrations of cel-miR-39-3p and argonaute RISC catalytic component 2 (AGO2) loading in ICs. This suggests the possibility of vertical transmission of miRNA signaling from milk through the neonatal digestive tract.
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-PLCXD3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-pan Ago Antibody, clone 2A8, clone 2A8, from mouse