MilliporeSigma
  • PRMT4 interacts with NCOA4 to inhibit ferritinophagy in cisplatin-induced acute kidney injury.
PRMT4 interacts with NCOA4 to inhibit ferritinophagy in cisplatin-induced acute kidney injury.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2024-04-03)
Lizhi Zhou, Zebin Deng, Yilong Wang, Hao Zhang, Shu Yan, Yashpal S Kanwar, Yinhuai Wang, Yingbo Dai, Fei Deng
ABSTRACT
Cisplatin-induced acute kidney injury (AKI) is commonly seen in the clinical practice, and ferroptosis, a type of non-apoptotic cell death, plays a pivotal role in it. Previous studies suggested that protein arginine methyltransferase 4 (PRMT4) was incorporated in various bioprocesses, but its role in renal injuries has not been investigated. Our present study showed that PRMT4 was highly expressed in renal proximal tubular cells, and it was downregulated in cisplatin-induced AKI. Besides, genetic disruption of PRMT4 exacerbated, while its overexpression attenuated, cisplatin-induced redox injuries in renal proximal epithelia. Mechanistically, our work showed that PRMT4 interacted with NCOA4 to inhibit ferritinophagy, a type of selective autophagy favoring lipid peroxidation to accelerate ferroptosis. Taken together, our study demonstrated that PRMT4 interacted with NCOA4 to attenuate ferroptosis in cisplatin-induced AKI, suggesting that PRMT4 might present as a new therapeutic target for cisplatin-related nephropathy.
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)