MilliporeSigma
  • Cell atlas of the regenerating human liver after portal vein embolization.
Cell atlas of the regenerating human liver after portal vein embolization.
Nature communications (2024-07-12)
Agnieska Brazovskaja, Tomás Gomes, Rene Holtackers, Philipp Wahle, Christiane Körner, Zhisong He, Theresa Schaffer, Julian Connor Eckel, René Hänsel, Malgorzata Santel, Makiko Seimiya, Timm Denecke, Michael Dannemann, Mario Brosch, Jochen Hampe, Daniel Seehofer, Georg Damm, J Gray Camp, Barbara Treutlein
ABSTRACT
The liver has the remarkable capacity to regenerate. In the clinic, regeneration is induced by portal vein embolization, which redirects portal blood flow, resulting in liver hypertrophy in locations with increased blood supply, and atrophy of embolized segments. Here, we apply single-cell and single-nucleus transcriptomics on healthy, hypertrophied, and atrophied patient-derived liver samples to explore cell states in the regenerating liver. Our data unveils pervasive upregulation of genes associated with developmental processes, cellular adhesion, and inflammation in post-portal vein embolization liver, disrupted portal-central hepatocyte zonation, and altered cell subtype composition of endothelial and immune cells. Interlineage crosstalk analysis reveals mesenchymal cells as an interaction hub between immune and endothelial cells, and highlights the importance of extracellular matrix proteins in liver regeneration. Moreover, we establish tissue-scale iterative indirect immunofluorescence imaging for high-dimensional spatial analysis of perivascular microenvironments, uncovering changes to tissue architecture in regenerating liver lobules. Altogether, our data is a rich resource revealing cellular and histological changes in human liver regeneration.
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid