MilliporeSigma
A direct estrogenic involvement in the expression of human hypocretin.
Life sciences (2024-03-22)
Haimei Li, Xinlu Chen, Jingyi Dong, Ripeng Liu, Jinfeng Duan, Manli Huang, Shaohua Hu, Jing Lu
ABSTRACT
Hypocretin is synthesized exclusively in the hypothalamus and distributes inputs to several areas of the brain, which may play an important role in depression. Our previous study showed that hypocretin-1 was increased in the lateral hypothalamus in female patients with depression compared to female controls. Estrogen acts through estrogen receptor (ER)α and ERβ. We studied the possibility of a direct action of estrogen receptors on the expression of human hypocretin. We found that hypocretin-1 plasma levels were significantly higher in female patients with depression than in female controls. Female depression estrogen receptors and hypocretin are colocalized in the human lateral hypothalamus, PC12, and SK-N-SH cells. The estrogen receptor response elements (ERE) that exist in the hypocretin promoter region may directly regulate the gene expression of hypocretin. The synchronicity of change of hypocretin and estradiol both in hypothalamus and plasma was verified in female rats. In the presence of estradiol, specific binding occurs between the recombinant human ER and hypocretin-ERE. Expression of ER combined with estradiol repressed hypocretin promoter activity via the ERE. In conclusion, we found that estradiol may directly affect hypocretin neurons in the human hypothalamus via ER binding to the hypocretin-ERE, which may lead to the sex-specific pathogenesis of depression.