MilliporeSigma
  • A simple and robust serum-free media for the proliferation of muscle cells.
A simple and robust serum-free media for the proliferation of muscle cells.
Food research international (Ottawa, Ont.) (2023-09-10)
Stig Skrivergaard, Jette Feveile Young, Navid Sahebekhtiari, Cameron Semper, Meenakshi Venkatesan, Alexei Savchenko, Peter J Stogios, Margrethe Therkildsen, Martin Krøyer Rasmussen
ABSTRACT
Cultivated meat production requires an efficient, robust and highly optimized serum-free cell culture media for the needed upscaling of muscle cell expansion. Existing formulations of serum-free media are complex, expensive and have not been optimized for muscle cells. Thus, we undertook this work to develop a simple and robust serum-free media for the proliferation of bovine satellite cells (SCs) through Design of Experiment (DOE) and Response Surface Methodology (RSM) using precise and high-throughput image-based cytometry. Proliferative attributes were investigated with transcriptomics and long-term performance was validated using multiple live assays. Here we formulated a media based on three highly optimized components; FGF2 (2 ng/mL), fetuin (600 µg/mL) and BSA (75 µg/mL) which together with an insulin-transferrin-selenium (1x) supplement, sustained the proliferation of bovine SCs, porcine SCs and murine C2C12 muscle cells. Remarkably, cells cultured in our media named Tri-basal 2.0+ performed better than cell cultured in 10% FBS, with respect to proliferation. Hence, the optimized Tri-basal 2.0+ enhanced serum-free cell attachment and long-term proliferation, providing an alternative solution to the use of FBS in the production of cultivated meat.
Sigma-Aldrich
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Myosin (Skeletal, Fast) antibody produced in mouse, clone MY-32, ascites fluid