MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • FABP4, GINS2 and CBX7 Expression in Cancer Cervix Tissues: Clinical, Pathological and Prognostic Implications.
FABP4, GINS2 and CBX7 Expression in Cancer Cervix Tissues: Clinical, Pathological and Prognostic Implications.
Iranian journal of pathology (2024-06-12)
Mariem A Elfeky, Rema H Faraj Saad, Mohamed Ali Alabiad, Mohammed Alorini, Rehab Hemeda, Ramadan M Ali, Loay M Gertallah, Mohamed Negm, Ahmed Mahmoud Abdou, Ahmed Baker A Alshaikh, Ahmed Elmaasrawy
ABSTRACT
Cervical cancer spreads to the pelvic lymph nodes, leading to a high incidence of cancer recurrence and unfavorable survival rates. Therefore, there is an urgent need to detect new predictive biomarkers for the early assessment of pelvic lymph node status in patients with cervical cancer. The current study aimed to assess the expression of FABP4, GINS2, and CBX7 in cervical cancer tissue to detect their prognostic and predictive roles in developing lymph node metastases in patients with that cancer type. We collected the tissues from patients with cervical cancer and evaluated the expression of FABP4, GINS2, and CBX7 using immunohistochemistry. We evaluated the association between their expression and clinicopathological and prognostic parameters. A high expression of FABP4 and GINS2 and a low expression of CBX7 were found to be positively associated with the old age group, large tumor size, high grade and lymphovascular involvement, para-uterine organ infiltration, advanced FIGO stage, chemotherapeutic resistance, and tumor recurrence. We demonstrated the oncogenic roles of FABP4 and GISN2 in addition to the on-co-suppressive roles of CBX7 in cervical cancer and their association with poor clinicopathological criteria and poor survival. Our results may indicate that FABP4, GISN2, and CBX7 could be considered predictive biomarkers of the occurrence of lymph node metastases in the cancer of the cervix preoperatively, which could be beneficial in the accurate preoperative design therapy.
Sigma-Aldrich
Anti-GINS2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution