MilliporeSigma
  • Methotrexate and sulforaphane loaded PBA-G5-PAMAM dendrimers as a combination therapy for anti-inflammatory response in an intra-articular joint arthritic animal model.
Methotrexate and sulforaphane loaded PBA-G5-PAMAM dendrimers as a combination therapy for anti-inflammatory response in an intra-articular joint arthritic animal model.
International journal of pharmaceutics (2023-06-20)
Edidiong Nkiruka Essien, Neeraja Revi, Vishal Khatri, Songyun Liu, Geoffrey Van Thiel, Divya Bijukumar
ABSTRACT
L-sulforaphane (LSF), a natural product developed from cruciferous vegetables, have shown potent anti-inflammatory effect in cancer as well as arthritis. However, the stable delivery of LSF remains a major challenge. Methotrexate (MTX) is currently the first line treatment for managing RA and is most effective in patients when used in combination with other anti-inflammatory or anti-rheumatic drugs. Here we developed phenylboronic acid-PAMAM dendrimer (PBA-G5D) nanoparticles conjugated MTX (MTX-PBA-G5D), and L-sulforaphane (LSF/PBA-G5D) loaded dendrimers. The MTX and LSF drug loading and release kinetics was analyzed using HPLC. The lipopolysaccharide (LPS) stimulated macrophages were treated with the formulations to study the inflammatory response in vitro. For in vivo studies, arthritis was induced in five-week-old female Wistar rats, and the MTX- and LSF/PBA-G5-D were injected via intra-articular injection for treatment and the arthritis reduction was scored by weight, knee diameter, and serum cytokine level measurement. The average size of the drug-nanoparticle conjugates ranged from 135 to 250 nm, with mostly cationic surface charges. The encapsulation efficiency of the drugs to the modified dendrimer was more than 60% with a slow release of drugs from the nanoparticles within 24 h at pH 7.4. Drugs in the nanoparticle formulation were biocompatible, with promising anti-inflammatory effects in vitro against LPS-activated murine macrophages. Further in vivo studies on arthritis induced female Wistar rats, revealed significant anti-arthritic effects based on the arthritic scoring from the knee diameter reading, and anti-inflammatory effects based on the serum cytokine levels. This study provides a promising strategy for utilizing PAMAM dendrimers as a nanocarrier for LSF delivery for RA therapy.
Sigma-Aldrich
NanoFabTx phenyl boronate functionalized PAMAM dendrimer, 20 ml/ml aqueous solution, for protein delivery