MilliporeSigma
  • Insulin growth factor-1 promotes the proliferation and osteogenic differentiation of bone marrow mesenchymal stem cells through the Wnt/β-catenin pathway.
Insulin growth factor-1 promotes the proliferation and osteogenic differentiation of bone marrow mesenchymal stem cells through the Wnt/β-catenin pathway.
Experimental and therapeutic medicine (2021-07-02)
Jing Feng, Zhiqiang Meng
ABSTRACT
Bone marrow mesenchymal stem cells (BMSCs) are stem cells that exist in bone marrow tissue and have osteogenic differentiation potential. Insulin growth factor-1 (IGF-1) plays a key role in the proliferation and osteogenic differentiation of BMSCs. However, the specific mechanism of IGF-1 in cell proliferation and osteogenic differentiation remains unclear. In the present study, BMSCs were transfected with lentivirus carrying the siRNA-Wnt3a gene, and the Wnt3a level in BMSCs was revealed to be reduced by western blotting, real-time quantitative polymerase chain reaction and immunofluorescence detection. Then, BMSCs were treated with 80 ng/ml IGF-1 in complete medium for 5 days. CCK-8 and cell cycle assays revealed that cell proliferation was significantly decreased in the siRNA-Wnt3a group than in the control group. The protein and mRNA levels of β-catenin and cyclin D1 were significantly downregulated in the siRNA-Wnt3a group compared with the control group. In addition, BMSCs were treated with IGF-1 in osteogenic differentiation medium for 7 and 21 days, and alkaline phosphatase staining and Alizarin Red staining demonstrated significantly reduced osteogenic differentiation ability in the siRNA-Wnt3a group compared with the control group. Furthermore, the protein and mRNA levels of β-catenin, RUNX2, and OPN were downregulated compared with the control group. Our findings revealed that IGF-1 promoted the proliferation and differentiation of BMSCs at least partially through the Wnt/β-catenin pathway. These findings provided new insight into the clinical treatment of bone disease.
Sigma-Aldrich
TissueFab® PODS Growth Factor Loaded Bioink Kit, Bone differentiation