MilliporeSigma
Reactive oxygen species enhance insulin sensitivity.
Cell metabolism (2009-10-08)
Kim Loh, Haiyang Deng, Atsushi Fukushima, Xiaochu Cai, Benoit Boivin, Sandra Galic, Clinton Bruce, Benjamin J Shields, Beata Skiba, Lisa M Ooms, Nigel Stepto, Ben Wu, Christina A Mitchell, Nicholas K Tonks, Matthew J Watt, Mark A Febbraio, Peter J Crack, Sofianos Andrikopoulos, Tony Tiganis
ABSTRACT
Chronic reactive oxygen species (ROS) production by mitochondria may contribute to the development of insulin resistance, a primary feature of type 2 diabetes. In recent years it has become apparent that ROS generation in response to physiological stimuli such as insulin may also facilitate signaling by reversibly oxidizing and inhibiting protein tyrosine phosphatases (PTPs). Here we report that mice lacking one of the key enzymes involved in the elimination of physiological ROS, glutathione peroxidase 1 (Gpx1), were protected from high-fat-diet-induced insulin resistance. The increased insulin sensitivity in Gpx1(-/-) mice was attributed to insulin-induced phosphatidylinositol-3-kinase/Akt signaling and glucose uptake in muscle and could be reversed by the antioxidant N-acetylcysteine. Increased insulin signaling correlated with enhanced oxidation of the PTP family member PTEN, which terminates signals generated by phosphatidylinositol-3-kinase. These studies provide causal evidence for the enhancement of insulin signaling by ROS in vivo.
Sigma-Aldrich
Horse Serum, Donor Herd, USA origin, Heat inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Horse Serum, Donor herd, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Horse serum, liquid
Sigma-Aldrich
Horse Serum, USA origin, Donor herd, suitable for cell culture, suitable for hybridoma