MilliporeSigma
Accidental acute fatal sodium nitrite poisoning.
Journal of toxicology. Clinical toxicology (1995-01-01)
S Gautami, R N Rao, T C Raghuram, S Rajagopalan, R V Bhat
ABSTRACT
An accidental food poisoning outbreak occurred resulting in the death of 14 of the 22 affected persons. Chemical analysis of food materials revealed the presence of sodium nitrite and potassium arsenate. Analysis of urine, gastric contents, liver and kidney, collected at the time of autopsy, confirmed the presence of these chemical substances in high concentrations. Epidemiological data indicated that the food poisoning occurred due to the accidental use of sodium nitrite and potassium arsenate instead of table salt in the preparation of tamarind (Tamarindus indica) soup.