MilliporeSigma
Poison hemlock-induced respiratory failure in a toddler.
Pediatric emergency care (2009-11-17)
Patrick L West, B Zane Horowitz, Marc T Montanaro, James N Lindsay
ABSTRACT
The ingestion of poison hemlock, or Conium maculatum, is described in a 2-year-old boy. He had the onset of abdominal pain and weakness after being fed C. maculatum picked by his sister from the roadside 2 hours earlier. He had a rapidly progressive muscular weakness and was intubated for respiratory failure. His symptoms completely resolved within 24 hours of the ingestion. Conium maculatum is a common weed that causes toxicity by its primary toxin, coniine, which stimulates nicotinic receptors and causes a syndrome of rapidly progressive muscle weakness and paralysis. We describe the course of a benign-appearing plant ingestion resulting in respiratory failure.