MilliporeSigma
  • Highly efficient generation of definitive endoderm lineage from human induced pluripotent stem cells.
Highly efficient generation of definitive endoderm lineage from human induced pluripotent stem cells.
Transplantation proceedings (2012-05-09)
K Sekine, T Takebe, Y Suzuki, A Kamiya, H Nakauchi, H Taniguchi
ABSTRACT
Although hepatocytes can be an option for liver transplantation, the shortage of donor organs continues to worsen. Since the development of induced pluripotent stem (iPS) cell technology, it is eagerly anticipated to produce functional elements from pluripotent stem cells. These functional cells differentiated from iPS cells could be used for transplantation, drug screening, and in vitro toxicology. Human iPS cells are maintained on Mitomycin C-treated mouse embryonic fibroblast layers in DMEM-Ham F12-based medium supplemented with Knockout Serum Replacement, nonessential amino acids, 2-mercaptoethanol, and Glutamax. Differentiation of human iPS cells into a definitive endodermal lineage was induced with PRMI 1640 medium supplemented with B27 and 100 ng/mL human activin A. Two B27 supplements were examined with and without insulin. Furthermore, the PI3 kinase inhibitor LY294002 was used to examine the effect of inhibiting insulin signaling. We established efficient induction of definitive endodermal differentiation from iPS cells. Quantitative analysis revealed efficient (93.03 ± 2.74%) differentiation of human iPS cells into definitive endoderm cells using B27 minus insulin. This protocol may contribute as a fundamental technique to promote human iPS studies to develop cellular sources for transplantation.
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)