MilliporeSigma
  • Characterization of alpha-2 adrenergic receptors in the OK cell, an opossum kidney cell line.
Characterization of alpha-2 adrenergic receptors in the OK cell, an opossum kidney cell line.
The Journal of pharmacology and experimental therapeutics (1988-02-01)
T J Murphy, D B Bylund
ABSTRACT
We have characterized alpha-2 adrenergic receptors in OK cells, an opossum kidney-derived cell line. In membrane saturation binding experiments, [3H]rauwolscine (Kd = 74 pM) was 3-fold more potent than [3H]yohimbine (Kd = 230 pM). Each labeled a single class of binding sites with densities of 135 and 124 fmol/mg of protein for [3H]rauwolscine and [3H]yohimbine, respectively. Inhibition of [3H]rauwolscine and [3H]yohimbine binding by several alpha adrenergic agonists and antagonists demonstrated the radioligands labeled an alpha-2 type adrenergic receptor with a pharmacological profile similar to the alpha-2B receptor subtype. The rank order of potency for antagonist inhibition of binding was yohimbine greater than prazosin = phentolamine greater than chlorpromazine = corynanthine, whereas the rank order of agonist potency was oxymetazoline = clonidine greater than or equal to UK-14,304 greater than or equal to (-)-epinephrine greater than (-)-norepinephrine. The oxymetazoline, clonidine and antagonist inhibition curves were routinely monophasic and modeled best as a single class of binding sites. For the other agonists, inhibition binding curves were biphasic with approximately 35% of the binding sites existing in a high affinity state. These curves were shifted to the right in the presence of 0.1 mM GTP, and in general modeled as a single class of binding sites. UK-14,304, (-)-epinephrine, (-)-norepinephrine and oxymetazoline attenuated parathyroid hormone-stimulated cyclic AMP production by up to 70% in whole cell monolayers in a dose-dependent manner via a pertussis toxin-sensitive mechanism. With the exception of oxymetazoline, this inhibition could be reversed with alpha adrenergic antagonists.(ABSTRACT TRUNCATED AT 250 WORDS)
Product Description
OK, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.