MilliporeSigma
  • CysDB: a human cysteine database based on experimental quantitative chemoproteomics.
CysDB: a human cysteine database based on experimental quantitative chemoproteomics.
Cell chemical biology (2023-04-30)
Lisa M Boatner, Maria F Palafox, Devin K Schweppe, Keriann M Backus
ABSTRACT
Cysteine chemoproteomics provides proteome-wide portraits of the ligandability or potential "druggability" for thousands of cysteine residues. Consequently, these studies are facilitating resources for closing the druggability gap, namely, achieving pharmacological manipulation of ∼96% of the human proteome that remains untargeted by U.S. Food and Drug Administration (FDA) approved small molecules. Recent interactive datasets have enabled users to interface more readily with cysteine chemoproteomics datasets. However, these resources remain limited to single studies and therefore do not provide a mechanism to perform cross-study analyses. Here we report CysDB as a curated community-wide repository of human cysteine chemoproteomics data derived from nine high-coverage studies. CysDB is publicly available at https://backuslab.shinyapps.io/cysdb/ and features measures of identification for 62,888 cysteines (24% of the cysteinome), as well as annotations of functionality, druggability, disease relevance, genetic variation, and structural features. Most importantly, we have designed CysDB to incorporate new datasets to further support the continued growth of the druggable cysteinome.
Product Description
Desthiobiotin-iodoacetamide, ≥98% (HPLC), powder, pan-cysteine profiling tag