MilliporeSigma
  • The selective peroxisome proliferator-activated receptor-delta agonist seladelpar reverses nonalcoholic steatohepatitis pathology by abrogating lipotoxicity in diabetic obese mice.
The selective peroxisome proliferator-activated receptor-delta agonist seladelpar reverses nonalcoholic steatohepatitis pathology by abrogating lipotoxicity in diabetic obese mice.
Hepatology communications (2018-02-07)
Fahrettin Haczeyni, Hans Wang, Vanessa Barn, Auvro R Mridha, Matthew M Yeh, W Geoffrey Haigh, George N Ioannou, Yun-Jung Choi, Charles A McWherter, Narcissus C-H Teoh, Geoffrey C Farrell
ABSTRACT
Lipotoxicity associated with insulin resistance is central to nonalcoholic steatohepatitis (NASH) pathogenesis. To date, only weight loss fully reverses NASH pathology, but mixed peroxisome proliferator-activated receptor-alpha/delta (PPAR-α/δ) agonists show some efficacy. Seladelpar (MBX-8025), a selective PPAR-δ agonist, improves atherogenic dyslipidemia. We therefore used this agent to test whether selective PPAR-δ activation can reverse hepatic lipotoxicity and NASH in an obese, dyslipidemic, and diabetic mouse model. From weaning, female Alms1 mutant (foz/foz) mice and wild-type littermates were fed an atherogenic diet for 16 weeks; groups (n = 8-12) were then randomized to receive MBX-8025 (10 mg/kg) or vehicle (1% methylcellulose) by gavage for 8 weeks. Despite minimally altering body weight, MBX-8025 normalized hyperglycemia, hyperinsulinemia, and glucose disposal in foz/foz mice. Serum alanine aminotransferase ranged 300-600 U/L in vehicle-treated foz/foz mice; MBX-8025 reduced alanine aminotransferase by 50%. In addition, MBX-8025 normalized serum lipids and hepatic levels of free cholesterol and other lipotoxic lipids that were increased in vehicle-treated foz/foz versus wild-type mice. This abolished hepatocyte ballooning and apoptosis, substantially reduced steatosis and liver inflammation, and improved liver fibrosis. In vehicle-treated foz/foz mice, the mean nonalcoholic fatty liver disease activity score was 6.9, indicating NASH; MBX-8025 reversed NASH in all foz/foz mice (nonalcoholic fatty liver disease activity score 3.13). Conclusion: Seladelpar improves insulin sensitivity and reverses dyslipidemia and hepatic storage of lipotoxic lipids to improve NASH pathology in atherogenic diet-fed obese diabetic mice. Selective PPAR-δ agonists act independently of weight reduction, but counter lipotoxicity related to insulin resistance, thereby providing a novel therapy for NASH. (Hepatology Communications 2017;1:663-674).
Product Description
Seladelpar, ≥98% (HPLC), (liquid, semi-solid, solid or powder), PPARδ agonist