MilliporeSigma
  • Cytotoxic effects of kinetin riboside and its selected analogues on cancer cell lines.
Cytotoxic effects of kinetin riboside and its selected analogues on cancer cell lines.
Bioorganic & medicinal chemistry letters (2024-01-28)
Ewa Totoń, Natalia Lisiak, Aleksandra Romaniuk-Drapała, Grzegorz Framski, Eliza Wyszko, Tomasz Ostrowski
ABSTRACT
N6-[(Furan-2-yl)methyl]adenosine (kinetin riboside) and its seven synthesized analogues were examined for the ability to inhibit the growth of five human carcinoma cell lines and for comparison of normal human lung fibroblast cell line (MRC-5). Out of the compounds evaluated, 8-azakinetin riboside was shown to exhibit significant cytotoxic activity for 72 h treatment against ovarian OVCAR-3 and pancreatic MIA PaCa-2 cancer cells (IC50 = 1.1 μM) with an observed weaker effect against MRC-5 cells (IC50 = 4.6 μM). Kinetin riboside, as well as its N6-[(furan-3-yl)methyl]- and N6-[(thien-2-yl)methyl]- counterparts, also exhibited cytotoxic activities at low micromolar levels but were non-selective over MRC-5 cells.
Product Description
N6-Furfuryladenosine, ≥98% (HPLC)