MilliporeSigma
  • A β-hydroxybutyrate shunt pathway generates anti-obesity ketone metabolites.
A β-hydroxybutyrate shunt pathway generates anti-obesity ketone metabolites.
Cell (2024-11-14)
Maria Dolores Moya-Garzon, Mengjie Wang, Veronica L Li, Xuchao Lyu, Wei Wei, Alan Sheng-Hwa Tung, Steffen H Raun, Meng Zhao, Laetitia Coassolo, Hashim Islam, Barbara Oliveira, Yuqin Dai, Jan Spaas, Antonio Delgado-Gonzalez, Kenyi Donoso, Aurora Alvarez-Buylla, Francisco Franco-Montalban, Anudari Letian, Catherine P Ward, Lichao Liu, Katrin J Svensson, Emily L Goldberg, Christopher D Gardner, Jonathan P Little, Steven M Banik, Yong Xu, Jonathan Z Long
ABSTRACT
β-Hydroxybutyrate (BHB) is an abundant ketone body. To date, all known pathways of BHB metabolism involve the interconversion of BHB and primary energy intermediates. Here, we identify a previously undescribed BHB secondary metabolic pathway via CNDP2-dependent enzymatic conjugation of BHB and free amino acids. This BHB shunt pathway generates a family of anti-obesity ketone metabolites, the BHB-amino acids. Genetic ablation of CNDP2 in mice eliminates tissue amino acid BHB-ylation activity and reduces BHB-amino acid levels. The most abundant BHB-amino acid, BHB-Phe, is a ketosis-inducible congener of Lac-Phe that activates hypothalamic and brainstem neurons and suppresses feeding. Conversely, CNDP2-KO mice exhibit increased food intake and body weight following exogenous ketone ester supplementation or a ketogenic diet. CNDP2-dependent amino acid BHB-ylation and BHB-amino acid metabolites are also conserved in humans. Therefore, enzymatic amino acid BHB-ylation defines a ketone shunt pathway and bioactive ketone metabolites linked to energy balance.
Product Description
BHB-Phe, ≥98% (HPLC), powder, BHB secondary metabolite