MilliporeSigma
  • A Hemilabile NHC-Gold Complex and its Application to the Redox Neutral 1,2-Oxyarylation of Feedstock Alkenes.
A Hemilabile NHC-Gold Complex and its Application to the Redox Neutral 1,2-Oxyarylation of Feedstock Alkenes.
Angewandte Chemie (International ed. in English) (2023-03-31)
Samuel C Scott, Jamie A Cadge, Grace K Boden, John F Bower, Christopher A Russell
ABSTRACT
We describe a AuI complex of a hemi-labile (C^N) N-heterocyclic carbene ligand that is able to mediate oxidative addition of aryl iodides. Detailed computational and experimental investigations have been undertaken to verify and rationalize the oxidative addition process. Application of this initiation mode has resulted in the first examples of "exogenous oxidant-free" AuI /AuIII catalyzed 1,2-oxyarylations of ethylene and propylene. These demanding yet powerful processes establish these commodity chemicals as nucleophilic-electrophilic building blocks in catalytic reaction design.
Sigma-Aldrich
RUMeIPr, ≥95%