MilliporeSigma
  • Artepillin C, a major component of Brazilian green propolis, inhibits endoplasmic reticulum stress and protein aggregation.
Artepillin C, a major component of Brazilian green propolis, inhibits endoplasmic reticulum stress and protein aggregation.
European journal of pharmacology (2021-10-18)
Yoko Hirata, Masanari Motoyama, Shintaro Kimura, Madoka Takashima, Takanori Ikawa, Kentaro Oh-Hashi, Yuji O Kamatari
ABSTRACT
Propolis, a compound produced by honeybees, has long been used in food and beverages to improve health and prevent diseases. We previously reported that the ethanol extracts of Brazilian green propolis and its constituents artepillin C, kaempferide, and kaempferol mitigate oxidative stress-induced cell death via oxytosis/ferroptosis. Here, we investigated the potential of Brazilian green propolis and its constituents to protect against endoplasmic reticulum stress in the mouse hippocampal cell line HT22. Ethanol extracts of Brazilian green propolis, artepillin C, and kaempferol attenuated tunicamycin-induced unfolded protein response and cell death. Interestingly, artepillin C inhibited both tunicamycin-induced protein aggregation in HT22 cells and the spontaneous protein aggregation of mutant canine superoxide dismutase 1 (E40K-SOD1-EGFP) in Neuro2a cells. These findings indicate that in addition to oxidative stress, the ethanol extracts of Brazilian green propolis help prevent endoplasmic reticulum stress-related neuronal cell death, which is proposedly involved in several neurodegenerative diseases. Moreover, artepillin C, a major constituent of Brazilian green propolis, may exhibit chemical chaperone-like properties.
Product Description
Artepillin C, ≥98% (HPLC), powder, CREB-CRTC2 inhibitor