- "Cationic" Suzuki-Miyaura Coupling with Acutely Base-Sensitive Boronic Acids.
"Cationic" Suzuki-Miyaura Coupling with Acutely Base-Sensitive Boronic Acids.
Journal of the American Chemical Society (2017-09-02)
Liye Chen, Daniel R Sanchez, Bufan Zhang, Brad P Carrow
PMID28862445
ABSTRACT
Fast, base-promoted protodeboronation of polyfluoroaryl and heteroaryl boronic acids complicates their use in Suzuki-Miyaura coupling (SMC) because a base is generally required for catalysis. We report a "cationic" SMC method using a PAd3-Pd catalyst that proceeds at rt in the absence of a base or metal mediator. A wide range of sensitive boronic acids, particularly polyfluoroaryl substrates that are poorly compatible with classic SMC conditions, undergo clean coupling. Stoichiometric experiments implicate the intermediacy of organopalladium cations, which supports a long-postulated cationic pathway for transmetalation in SMC.
MATERIALS