MilliporeSigma
  • Efficient and rapid template-directed nucleic acid copying using 2'-amino-2',3'-dideoxyribonucleoside-5'-phosphorimidazolide monomers.
Efficient and rapid template-directed nucleic acid copying using 2'-amino-2',3'-dideoxyribonucleoside-5'-phosphorimidazolide monomers.
Journal of the American Chemical Society (2009-09-18)
Jason P Schrum, Alonso Ricardo, Mathangi Krishnamurthy, J Craig Blain, Jack W Szostak
ABSTRACT
The development of a sequence-general nucleic acid copying system is an essential step in the assembly of a synthetic protocell, an autonomously replicating spatially localized chemical system capable of spontaneous Darwinian evolution. Previously described nonenzymatic template-copying experiments have validated the concept of nonenzymatic replication, but have not yet achieved robust, sequence-general polynucleotide replication. The 5'-phosphorimidazolides of the 2'-amino-2',3'-dideoxyribonucleotides are attractive as potential monomers for such a system because they polymerize by forming 2'-->5' linkages, which are favored in nonenzymatic polymerization reactions using similarly activated ribonucleotides on RNA templates. Furthermore, the 5'-activated 2'-amino nucleotides do not cyclize. We recently described the rapid and efficient nonenzymatic copying of a DNA homopolymer template (dC(15)) encapsulated within fatty acid vesicles using 2'-amino-2',3'-dideoxyguanosine-5'-phosphorimidazolide as the activated monomer. However, to realize a true Darwinian system, the template-copying chemistry must be able to copy most sequences and their complements to allow for the transmission of information from generation to generation. Here, we describe the copying of a series of nucleic acid templates using 2'-amino-2',3'-dideoxynucleotide-5'-phosphorimidazolides. Polymerization reactions proceed rapidly to completion on short homopolymer RNA and LNA templates, which favor an A-type duplex geometry. We show that more efficiently copied sequences are generated by replacing the adenine nucleobase with diaminopurine, and uracil with C5-(1-propynyl)uracil. Finally, we explore the copying of longer, mixed-sequence RNA templates to assess the sequence-general copying ability of 2'-amino-2',3'-dideoxynucleoside-5'-phosphorimidazolides. Our results are a significant step forward in the realization of a self-replicating genetic polymer compatible with protocell template copying and suggest that N2'-->P5'-phosphoramidate DNA may have the potential to function as a self-replicating system.