MilliporeSigma
  • E(y)2/Sus1 is required for blocking PRE silencing by the Wari insulator in Drosophila melanogaster.
E(y)2/Sus1 is required for blocking PRE silencing by the Wari insulator in Drosophila melanogaster.
Chromosoma (2010-01-19)
Maksim Erokhin, Alexander Parshikov, Pavel Georgiev, Darya Chetverina
ABSTRACT
Chromatin insulators affect interactions between promoters and enhancers/silencers and function as barriers to the spread of repressive chromatin. Recently, we have found an insulator, named Wari, located on the 3' side of the white gene. Here, we show that the previously identified 368-bp core of this insulator is sufficient for blocking Polycomb response element-mediated silencing. Although Wari does not contain binding sites for known insulator proteins, the E(y)2 and CP190 proteins bind to Wari as well as to the Su(Hw)-containing insulators in vivo. It may well be that these proteins are recruited to the insulator by as yet unidentified DNA-binding protein. Partial inactivation of E(y)2 in a weak e(y)2 ( u1 ) mutation impairs only the anti-silencing but not the enhancer-blocking activity of the Wari insulator. Thus, the E(y)2 protein in different Drosophila insulators serves to protect gene expression from silencing.
Sigma-Aldrich
IgG from rabbit serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Millipore
Protein G Sepharose, Fast Flow, recombinant, expressed in E. coli, aqueous ethanol suspension