MilliporeSigma
  • Vascular endothelial growth factor B, a novel growth factor for endothelial cells.
Vascular endothelial growth factor B, a novel growth factor for endothelial cells.
Proceedings of the National Academy of Sciences of the United States of America (1996-03-19)
B Olofsson, K Pajusola, A Kaipainen, G von Euler, V Joukov, O Saksela, A Orpana, R F Pettersson, K Alitalo, U Eriksson
ABSTRACT
We have isolated and characterized a novel growth factor for endothelial cells, vascular endothelial growth factor B (VEGF-B), with structural similarities to vascular endothelial growth factor (VEGF) and placenta growth factor. VEGF-B was particularly abundant in heart and skeletal muscle and was coexpressed with VEGF in these and other tissues. VEGF-B formed cell-surface-associated disulfide-linked homodimers and heterodimerized with VEGF when coexpressed. Conditioned medium from transfected 293EBNA cells expressing VEGF-B stimulated DNA synthesis in endothelial cells. Our results suggest that VEGF-B has a role in angiogenesis and endothelial cell growth, particularly in muscle.