MilliporeSigma
Activins and activin receptors in cell growth.
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.) (1997-02-01)
S Y Ying, Z Zhang, B Furst, Y Batres, G Huang, G Li
ABSTRACT
Activin and inhibin, members of transforming growth factor-beta (TGFbeta) superfamily, have diverse and widespread effects within living organisms at many stages during growth and development. From the initial isolation of these growth factors based on their effects of FSH secretion, the study of these factors, as well as of the activin-binding protein follistatin, has progressed from the localization of the expression of the inhibin alpha subunit, activin betaA and betaB subunits, and activin receptors in the tissues of various organisms to the examination of activin and inhibin as autocrine and paracrine agents in cell proliferation and differentiation. The inhibitory effects on cell growth and differentiation that have been observed upon treatment of cells with activin suggest that further understanding of the bioactivity of this molecule and its characterization on a molecular level may aid in a more complete understanding of cell growth and differentiation. This minireview discusses the roles of activin, inhibin, and follistatin in the arenas of cell proliferation, differentiation, and embryogenesis, as well as the roles of these molecules in cancerous cells.