MilliporeSigma
  • Rapid, dynamic changes in glomerular permeability to macromolecules during systemic angiotensin II (ANG II) infusion in rats.
Rapid, dynamic changes in glomerular permeability to macromolecules during systemic angiotensin II (ANG II) infusion in rats.
American Journal of Physiology: Renal Physiology (2012)
Axelsson J, Rippe A, Oberg CM, Rippe B.