MilliporeSigma
  • Purification and characterization of a newly identified growth factor specific for epithelial cells.
Purification and characterization of a newly identified growth factor specific for epithelial cells.
Proceedings of the National Academy of Sciences of the United States of America (1989-02-01)
J S Rubin, H Osada, P W Finch, W G Taylor, S Rudikoff, S A Aaronson
ABSTRACT
A growth factor specific for epithelial cells was identified in conditioned medium of a human embryonic lung fibroblast cell line. The factor, provisionally termed keratinocyte growth factor (KGF) because of its predominant activity on this cell type, was purified to homogeneity by a combination of ultrafiltration, heparin-Sepharose affinity chromatography, and hydrophobic chromatography on a C4 reversed-phase HPLC column. KGF was both acid and heat labile and consisted of a single polypeptide chain of approximately 28 kDa. Purified KGF was a potent mitogen for epithelial cells, capable of stimulating DNA synthesis in quiescent BALB/MK epidermal keratinocytes by greater than 500-fold with activity detectable at 0.1 nM and maximal at 1.0 nM. Lack of mitogenic activity on either fibroblasts or endothelial cells indicated that KGF possessed a target cell specificity distinct from any previously characterized growth factor. Microsequencing revealed an amino-terminal sequence containing no significant homology to any known protein. The release of this growth factor by human embryonic fibroblasts raises the possibility that KGF may play a role in mesenchymal stimulation of normal epithelial cell proliferation.
Sigma-Aldrich
Fibroblast Growth Factor-10 human, >97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Keratinocyte Growth Factor human, KGF, recombinant, expressed in E. coli, powder, suitable for cell culture