MilliporeSigma
  • A novel role of follistatin, an activin-binding protein, in the inhibition of activin action in rat pituitary cells. Endocytotic degradation of activin and its acceleration by follistatin associated with cell-surface heparan sulfate.
A novel role of follistatin, an activin-binding protein, in the inhibition of activin action in rat pituitary cells. Endocytotic degradation of activin and its acceleration by follistatin associated with cell-surface heparan sulfate.
The Journal of biological chemistry (1997-05-23)
O Hashimoto, T Nakamura, H Shoji, S Shimasaki, Y Hayashi, H Sugino
ABSTRACT
There are two types of the activin-binding protein follistatin (FS), FS-288 and FS-315. These result from alternative splicing of mRNA. FS-288 exhibits high affinity for cell-surface heparan sulfate proteoglycans, whereas FS-315 shows low affinity. To understand the physiological role of cell-associated FS, we investigated the binding of activin to cell-associated FS and its behavior on the cell surface using primary cultured rat pituitary cells. Affinity cross-linking experiments using 125I-activin A demonstrated that activin bound to rat pituitary cells via FS as well as to their receptors on the cell surface. FS-288 promoted the binding of activin A to the cell surface more markedly than FS-315. When the cells were incubated with 125I-activin A in the presence of FS-288, significant degradation of activin A was observed, and this was dependent on the FS-288 concentration. This activin degradation was abolished by heparan sulfate, chloroquine, and several lysosomal enzyme inhibitors. Moreover, FS-288 stimulated cellular uptake of activin A, whereas chloroquine suppressed lysosomal degradation following internalization, as demonstrated by microscopic autoradiography. These results suggest that cell-associated FS-288 accelerates the uptake of activin A into pituitary cells, leading to increased degradation by lysosomal enzymes, and thus plays a role in the activin clearance system.
Sigma-Aldrich
Follistatin 300 human, ≥90% (SDS-PAGE), recombinant, expressed in Sf21 cells, lyophilized powder, suitable for cell culture
Pricing and availability is not currently available.