MilliporeSigma
  • The cleavage site for the restriction endonuclease EcoRV is 5'-GAT/ATC-3'.
The cleavage site for the restriction endonuclease EcoRV is 5'-GAT/ATC-3'.
Gene (1984-03-01)
I Schildkraut, C D Banner, C S Rhodes, S Parekh
ABSTRACT
The cleavage site for the restriction endonuclease EcoRV has been found to be 5'-GAT/ATC-3', rather than 5'- GATAT /C-3' as reported earlier by Kholmina et al. [ Dokl . Akad . Nauk . 253 (1980) 495-497].