MilliporeSigma
  • Regulation of gene expression by low levels of ultraviolet-B radiation in Pisum sativum: isolation of novel genes by suppression subtractive hybridisation.
Regulation of gene expression by low levels of ultraviolet-B radiation in Pisum sativum: isolation of novel genes by suppression subtractive hybridisation.
Plant & cell physiology (2002-04-30)
Helena Sävenstrand, Mikael Brosché, Ake Strid
ABSTRACT
Suppression subtractive hybridisation was used to isolate genes differentially regulated by low levels (UV-B(BE,300) 0.13 W m(-2)) of ultraviolet-B radiation (UV-B; 290-320 nm) in Pisum sativum. Six genes were regulated, two of which were novel. The mRNA levels for these two (PsTSDC and PsUOS1) were increased and depressed by UV-B treatment, respectively. Domains in the PsTSDC translation product was similar to TIR (Toll-Interleukin-1 receptor-similar) domains and a NB-ARC domain (nucleotide-binding domain in APAF-1, R gene products and CED-4). The PsUOS1 translation product was similar to an open reading frame in Arabidopsis. Genes encoding embryo-abundant protein (PsEMB) and S-adenosyl-L-methionine synthase (PsSAMS) were induced by UV-B, whereas the transcript levels for genes encoding sucrose transport protein (PsSUT) or ribulose-5-phosphate 3-epimerase (PsR5P3E) were decreased. These regulation patterns are novel, and the PsEMB and PsR5P3E sequences are reported for the first time. The stress-specificity of regulation of these genes were tested by ozone fumigation (100 ppb O(3)). Qualitatively, the similarity of expression after both UV-B and ozone exposure suggests that, for these genes, similar stress-response pathways are in action.