MilliporeSigma
  • In silico exploration of the fructose-6-phosphate phosphorylation step in glycolysis: genomic evidence of the coexistence of an atypical ATP-dependent along with a PPi-dependent phosphofructokinase in Propionibacterium freudenreichii subsp. shermanii.
In silico exploration of the fructose-6-phosphate phosphorylation step in glycolysis: genomic evidence of the coexistence of an atypical ATP-dependent along with a PPi-dependent phosphofructokinase in Propionibacterium freudenreichii subsp. shermanii.
In silico biology (2004-10-28)
Guillaume Meurice, Catherine Deborde, Daniel Jacob, Hélène Falentin, Patrick Boyaval, Diliana Dimova
ABSTRACT
We performed a detailed bioinformatic study of the catalytic step of fructose-6-phosphate phosphorylation in glycolysis based on the raw genomic draft of Propionibacterium freudenreichii subsp. shermanii (P. shermanii) ATCC9614 [Meurice et al., 2004]. Our results provide the first in silico evidence of the coexistence of genes coding for an ATP-dependent phosphofructokinase (ATP-PFK) and a PPi-dependent phosphofructokinase (PPi-PFK), whereas the fructose-1,6-bisphosphatase (FBP) and ADP-dependent phosphofructokinase (ADP-PFK) are absent. The deduced amino acid sequence corresponding to the PPi-PFK (AJ508922) shares 100% similarity with the already characterised propionibacterial protein (P29495; Ladror et al., 1991]. The unexpected ATP-PFK gene (AJ509827) encodes a protein of 373 aa which is highly similar (50% positive residues) along at least 95% of its sequence length to different well-characterised ATP-PFKs. The characteristic PROSITE pattern important for the enzyme function of ATP-PFKs (PS00433) was conserved in the putative ATP-PFK sequence: 8 out of 9 amino acid residues. According to the recent evolutionary study of PFK proteins with different phosphate donors [Bapteste et al., 2003], the propionibacterial ATP-PFK harbours a G104-K124 residue combination, which strongly suggested that this enzyme belongs to the group of atypical ATP-PFKs. According to our phylogenetic analyses the amino acid sequence of the ATP-PFK is clustered with the atypical ATP-PFKs from group III of the Siebers classification [Siebers et al., 1998], whereas the expected PPi-PFK protein is closer to the PPi-PFKs from clade P [Müller et al., 2001]. The possible significance of the co-existence of these two PFKs and their importance for the regulation of glycolytic pathway flux in P. shermanii is discussed.