MilliporeSigma
  • Structure of a ribulose 5-phosphate 3-epimerase from Plasmodium falciparum.
Structure of a ribulose 5-phosphate 3-epimerase from Plasmodium falciparum.
Proteins (2005-11-24)
J Caruthers, J Bosch, F Buckner, W Van Voorhis, P Myler, E Worthey, C Mehlin, E Boni, G DeTitta, J Luft, A Lauricella, O Kalyuzhniy, L Anderson, F Zucker, M Soltis, Wim G J Hol
ABSTRACT
The crystal structure of Pfal009167AAA, a putative ribulose 5-phosphate 3-epimerase (PfalRPE) from Plasmodium falciparum, has been determined to 2 A resolution. RPE represents an exciting potential drug target for developing antimalarials because it is involved in the shikimate and the pentose phosphate pathways. The structure is a classic TIM-barrel fold. A coordinated Zn ion and a bound sulfate ion in the active site of the enzyme allow for a greater understanding of the mechanism of action of this enzyme. This structure is solved in the framework of the Structural Genomics of Pathogenic Protozoa (SGPP) consortium.