MilliporeSigma
  • Stereoselective biocatalytic synthesis of (S)-2-hydroxy-2-methylbutyric acid via substrate engineering by using "thio-disguised" precursors and oxynitrilase catalysis.
Stereoselective biocatalytic synthesis of (S)-2-hydroxy-2-methylbutyric acid via substrate engineering by using "thio-disguised" precursors and oxynitrilase catalysis.
Chemistry (Weinheim an der Bergstrasse, Germany) (2007-01-18)
Martin H Fechter, Karl Gruber, Manuela Avi, Wolfgang Skranc, Christian Schuster, Peter Pöchlauer, Kurt O Klepp, Herfried Griengl
ABSTRACT
3-Tetrahydrothiophenone (4) and 4-phenylthiobutan-2-one (7) were used as masked 2-butanone equivalents to give the corresponding cyanohydrins 5 (79 % yield, 91 % ee) and 8 (95 % yield, 96 % ee) in an enzymatic cyanohydrin reaction applying the hydroxynitrile lyase (HNL) from Hevea brasiliensis. After hydrolysis and desulphurisation the desired intermediate (S)-2-hydroxy-2-methylbutyric acid (10) was obtained with 99 % ee. Interestingly, when applying (R)-selective HNL from Prunus amygdalus again the (S)-cyanohydrin 5 was formed (62 % ee). The absolute configuration of 5 was verified by crystal structure determination of the corresponding hydrolysis derived carboxylate. The fact that both enzymes yield the same enantiomer was analysed and interpreted by molecular modelling calculations.